Assay Details
Target Gene Details
Entrez Gene ID: | 55246 |
Gene Name: | coiled-coil domain containing 25 |
Gene Aliases: |
- |
Location: |
Chr.8:27733316-27772656 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC25 | NM_001304529.1 | 7 | 879 | NP_001291458.1 |
| NM_001304530.1 | 7 | 1088 | NP_001291459.1 | |
| NM_001304532.1 | 8 | 1134 | NP_001291461.1 | |
| NM_018246.2 | 9 | 1186 | NP_060716.2 | |
| NR_130761.1 | 7 | 1036 | ||
| NR_130762.1 | 7 | 1030 | ||
| XM_011544570.2 | 8 | 1133 | XP_011542872.1 | |
| XM_011544571.2 | 7 | 1012 | XP_011542873.1 | |
| AK001715.1 | 8 | 1134 | BAA91857.1 | |
| AK027900.1 | 9 | 1138 | BAB55442.1 | |
| AK311437.1 | ||||
| BC032588.1 | 8 | 1123 | ||
| BC049197.1 | 9 | 1140 | ||
| BC065295.1 | 9 | 1127 | AAH65295.1 | |
| BC110391.1 | 7 | 971 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv981922 | Chr.8:27734880 - 27735880 on Build GRCh38 | Duplication |
|
| nsv610867 | Chr.8:27607002 - 27757379 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113447699] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map