Assay Details
Target Gene Details
Entrez Gene ID: | 5288 |
Gene Name: | phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma |
Gene Aliases: |
PI3K-C2-gamma, PI3K-C2GAMMA |
Location: |
Chr.12:18242946-18716861 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3C2G | NM_001288772.1 | 33 | 4735 | NP_001275701.1 |
| NM_001288774.1 | 33 | 4728 | NP_001275703.1 | |
| NM_004570.5 | 32 | 4612 | NP_004561.3 | |
| XM_011520696.2 | XP_011518998.1 | |||
| XM_011520697.2 | XP_011518999.1 | |||
| XM_017019470.1 | XP_016874959.1 | |||
| XM_017019471.1 | XP_016874960.1 | |||
| XM_017019472.1 | XP_016874961.1 | |||
| XM_017019473.1 | XP_016874962.1 | |||
| XM_017019474.1 | XP_016874963.1 | |||
| XM_017019476.1 | XP_016874965.1 | |||
| XM_017019477.1 | XP_016874966.1 | |||
| XM_017019479.1 | XP_016874968.1 | |||
| AI478164.1 | 1 | 237 | ||
| AJ000008.1 | 32 | 4634 | CAA03853.1 |
Target Gene Details
Entrez Gene ID: | 89869 |
Gene Name: | phospholipase C zeta 1 |
Gene Aliases: |
NYD-SP27, PLC-zeta-1, PLCzeta |
Location: |
Chr.12:18645529-18738059 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCZ1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv628 | Chr.12:18633645 - 18666360 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map