Assay Details
Target Gene Details
Entrez Gene ID: | 58484 |
Gene Name: | NLR family CARD domain containing 4 |
Gene Aliases: |
AIFEC, CARD12, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, CLR2.1, FCAS4, IPAF |
Location: |
Chr.2:32224449-32265743 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRC4 | NM_001199138.1 | 9 | 3258 | NP_001186067.1 |
| NM_001199139.1 | 9 | 3141 | NP_001186068.1 | |
| NM_001302504.1 | 8 | 1277 | NP_001289433.1 | |
| NM_021209.4 | 9 | 3272 | NP_067032.3 | |
| AF376061.1 | 10 | 3486 | AAK53443.1 | |
| AK095467.1 | 9 | 3258 | ||
| AK292673.1 | 9 | 3243 | ||
| AK314762.1 | 9 | 3243 | ||
| AL389934.1 | 6 | 1243 | CAB97523.1 | |
| AW337918.1 | 1 | 114 | ||
| AY027787.1 | 9 | 3273 | AAK14776.1 | |
| AY027788.1 | 8 | 1278 | AAK14777.1 | |
| AY032589.1 | 8 | 3032 | AAK38730.1 | |
| AY035391.1 | 9 | 3141 | AAK59843.1 | |
| AY358152.1 | 9 | 3220 | AAQ88519.1 | |
| BC031555.1 | 9 | 3228 | AAH31555.1 |
Target Gene Details
Entrez Gene ID: | 55676 |
Gene Name: | solute carrier family 30 member 6 |
Gene Aliases: |
MST103, MSTP103, ZNT6 |
Location: |
Chr.2:32165841-32224806 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC30A6 | XM_006712048.3 | 11 | 5066 | XP_006712111.1 |
| XM_011532959.2 | 14 | 5312 | XP_011531261.1 | |
| XM_011532960.2 | 14 | 5540 | XP_011531262.1 | |
| XM_011532961.2 | 13 | 5218 | XP_011531263.1 | |
| XM_011532962.2 | 12 | 5369 | XP_011531264.1 | |
| XM_017004462.1 | 12 | 5131 | XP_016859951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3590243 | Chr.2:32217953 - 32258270 on Build GRCh38 | Gain |
|
| nsv2667 | Chr.2:32219741 - 32264686 on Build GRCh38 | Deletion |
|
| esv2422335 | Chr.2:32187261 - 32975514 on Build GRCh38 | Duplication |
|
| nsv833670 | Chr.2:32070709 - 32256788 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map