Assay Details
Target Gene Details
Entrez Gene ID: | 7203 |
Gene Name: | chaperonin containing TCP1 subunit 3 |
Gene Aliases: |
CCT-gamma, CCTG, PIG48, TCP-1-gamma, TRIC5 |
Location: |
Chr.1:156308961-156338415 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT3 | NM_001008800.2 | NP_001008800.1 | ||
| NM_005998.4 | NP_005989.3 | |||
| NR_036564.1 | ||||
| NR_036565.1 | ||||
| AB208882.1 | BAD92119.1 | |||
| AK126400.1 | ||||
| AK293477.1 | ||||
| AK300765.1 | ||||
| AK315923.1 | ||||
| AK316387.1 | ||||
| AL833197.1 | CAI46192.1 | |||
| AY524046.1 | AAS94255.1 | |||
| BC006501.2 | ||||
| BC008019.1 | AAH08019.1 | |||
| U17104.1 | AAC50068.1 | |||
| X74801.1 | CAA52808.1 |
More Information
Set Membership: |
|

Genomic Map