Assay Details
Target Gene Details
Entrez Gene ID: | 7167 |
Gene Name: | triosephosphate isomerase 1 |
Gene Aliases: |
HEL-S-49, TIM, TPI, TPID |
Location: |
Chr.12:6867420-6870946 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPI1 | NM_000365.5 | NP_000356.1 | ||
| NM_001159287.1 | NP_001152759.1 | |||
| NM_001258026.1 | NP_001244955.1 | |||
| AK129488.1 | ||||
| AK222638.1 | BAD96358.1 | |||
| AK298809.1 | ||||
| AK300661.1 | ||||
| AK313282.1 | ||||
| BC007086.1 | AAH07086.1 | |||
| BC007812.2 | AAH07812.1 | |||
| BC009329.2 | AAH09329.1 | |||
| BC011611.2 | AAH11611.1 | |||
| BC015100.1 | AAH15100.1 | |||
| BC017165.1 | AAH17165.1 | |||
| BC017917.1 | AAH17917.1 | |||
| CR541702.1 | CAG46503.1 | |||
| EU794668.1 | ||||
| M10036.1 | AAB59511.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832324 | Chr.12:6780297 - 6932306 on Build GRCh38 | Loss |
|
| nsv557261 | Chr.12:6839304 - 6924660 on Build GRCh38 | Loss |
|
| esv3580405 | Chr.12:6826466 - 6903969 on Build GRCh38 | Gain |
|
| nsv983267 | Chr.12:6869188 - 6870599 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map