Assay Details
Target Gene Details
Entrez Gene ID: | 54492 |
Gene Name: | neuralized E3 ubiquitin protein ligase 1B |
Gene Aliases: |
NEURL3, hNeur2, neur2 |
Location: |
Chr.5:172641266-172691540 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEURL1B | NM_001142651.2 | NP_001136123.1 | ||
| NM_001308177.1 | NP_001295106.1 | |||
| NM_001308178.1 | NP_001295107.1 | |||
| GQ414757.1 | ||||
| GQ414758.1 | ||||
| GQ414759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600287 | Chr.5:172627918 - 172694311 on Build GRCh38 | Loss |
|
| esv34432 | Chr.5:172591201 - 172752581 on Build GRCh38 | Gain |
|
| nsv462526 | Chr.5:172616312 - 172652886 on Build GRCh38 | Loss |
|
| esv2759393 | Chr.5:172554329 - 172788409 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map