Assay Details
Target Gene Details
Entrez Gene ID: | 54492 |
Gene Name: | neuralized E3 ubiquitin protein ligase 1B |
Gene Aliases: |
NEURL3, hNeur2, neur2 |
Location: |
Chr.5:172641266-172691540 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEURL1B | NM_001142651.2 | NP_001136123.1 | ||
| NM_001308177.1 | NP_001295106.1 | |||
| NM_001308178.1 | NP_001295107.1 | |||
| GQ414757.1 | ||||
| GQ414758.1 | ||||
| GQ414759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv208e55 | Chr.5:172656276 - 172752581 on Build GRCh38 | Gain |
|
| nsv600289 | Chr.5:172657047 - 172700301 on Build GRCh38 | Loss |
|
| nsv1032497 | Chr.5:172660175 - 172726439 on Build GRCh38 | Gain |
|
| esv3607596 | Chr.5:172660394 - 172725776 on Build GRCh38 | Gain |
|
| nsv600287 | Chr.5:172627918 - 172694311 on Build GRCh38 | Loss |
|
| esv34432 | Chr.5:172591201 - 172752581 on Build GRCh38 | Gain |
|
| dgv5831n100 | Chr.5:172645828 - 172753954 on Build GRCh38 | Gain |
|
| esv2759393 | Chr.5:172554329 - 172788409 on Build GRCh38 | Gain |
|
| esv2763518 | Chr.5:172653533 - 172735933 on Build GRCh38 | Gain |
|
| nsv523045 | Chr.5:172652886 - 172728004 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs532094963] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map