Assay Details
Target Gene Details
Entrez Gene ID: | 22996 |
Gene Name: | tetratricopeptide repeat domain 39A |
Gene Aliases: |
C1orf34, DEME-6 |
Location: |
Chr.1:51287258-51345116 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC39A | NM_001080494.3 | 18 | 2194 | NP_001073963.1 |
| NM_001144832.2 | 18 | 2169 | NP_001138304.1 | |
| NM_001297663.1 | 18 | 2171 | NP_001284592.1 | |
| NM_001297664.1 | 18 | 2207 | NP_001284593.1 | |
| NM_001297665.1 | 18 | 2299 | NP_001284594.1 | |
| NM_001297666.1 | 7 | 1202 | NP_001284595.1 | |
| NM_001297667.1 | 6 | 1110 | NP_001284596.1 | |
| XM_005270644.2 | 18 | 2200 | XP_005270701.1 | |
| XM_006710471.2 | 18 | 2297 | XP_006710534.1 | |
| XM_011541048.2 | 18 | 2274 | XP_011539350.1 | |
| XM_011541049.2 | 18 | 2413 | XP_011539351.1 | |
| XM_011541050.2 | 18 | 2274 | XP_011539352.1 | |
| XM_011541051.2 | 18 | 2307 | XP_011539353.1 | |
| XM_011541052.2 | 18 | 2253 | XP_011539354.1 | |
| XM_011541053.2 | 18 | 2291 | XP_011539355.1 | |
| XM_011541054.2 | 18 | 2127 | XP_011539356.1 | |
| XM_017000715.1 | 18 | 2174 | XP_016856204.1 | |
| AF007170.1 | 18 | 2075 | AAC39582.1 | |
| AK298019.1 | ||||
| AK301587.1 | ||||
| BC004399.1 | 7 | 1163 | AAH04399.1 | |
| BC028374.2 | 18 | 2241 | AAH28374.1 |
More Information
Set Membership: |
|

Genomic Map