Assay Details
Target Gene Details
Entrez Gene ID: | 23358 |
Gene Name: | ubiquitin specific peptidase 24 |
Gene Aliases: |
- |
Location: |
Chr.1:55066359-55215374 on Build GRCh38 |
Assay Gene Location: | Within Intron 71 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP24 | NM_015306.2 | NP_056121.2 | ||
| XM_005270690.3 | XP_005270747.1 | |||
| XM_017000831.1 | XP_016856320.1 | |||
| XM_017000832.1 | XP_016856321.1 | |||
| XM_017000833.1 | XP_016856322.1 | |||
| XM_017000834.1 | XP_016856323.1 | |||
| AB028980.1 | BAA83009.1 | |||
| AK000321.1 | BAA91084.1 | |||
| AL832370.1 | ||||
| BC029660.1 | AAH29660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518539 | Chr.1:55034756 - 55087881 on Build GRCh38 | Loss |
|
| esv3577877 | Chr.1:55068894 - 55073692 on Build GRCh38 | Loss |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map