Assay Details
Target Gene Details
Entrez Gene ID: | 253152 |
Gene Name: | epoxide hydrolase 4 |
Gene Aliases: |
ABHD7, EH4, EPHXRP |
Location: |
Chr.1:92029976-92063536 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHX4 | NM_173567.4 | NP_775838.3 | ||
| AK074822.1 | BAC11230.1 | |||
| BC041475.1 | AAH41475.1 | |||
| DB475492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv365n54 | Chr.1:91750168 - 92133148 on Build GRCh38 | Gain |
|
| nsv1002730 | Chr.1:91836984 - 92099672 on Build GRCh38 | Gain |
|
| nsv830581 | Chr.1:91851650 - 92045618 on Build GRCh38 | Loss |
|
| esv2750842 | Chr.1:91891224 - 92114118 on Build GRCh38 | Gain |
|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
| nsv1006318 | Chr.1:91750851 - 92142767 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77406616] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map