Assay Details
Target Gene Details
Entrez Gene ID: | 730102 |
Gene Name: | quinone oxidoreductase-like protein 2 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.1:178006136-178038007 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC730102 | NR_037167.1 | |||
| BX538051.1 | ||||
| DA045540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548252 | Chr.1:177758922 - 178428784 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75949862] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map