Assay Details
Target Gene Details
Entrez Gene ID: | 1131 |
Gene Name: | cholinergic receptor muscarinic 3 |
Gene Aliases: |
EGBRS, HM3, PBS |
Location: |
Chr.1:239386565-239915450 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Gene Details
Entrez Gene ID: | 100506915 |
Gene Name: | CHRM3 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.1:239707126-239719119 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRM3-AS2 | NR_103776.1 | |||
| AL832284.1 | ||||
| CT002712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549435 | Chr.1:239211710 - 239810706 on Build GRCh38 | Gain |
|
| nsv954002 | Chr.1:239714601 - 239734400 on Build GRCh38 | Duplication |
|
| esv2759009 | Chr.1:239602794 - 240226204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map