Assay Details
Target Gene Details
Entrez Gene ID: | 728463 |
Gene Name: | TGFB2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.1:218344196-218345678 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB2-AS1 | NR_046268.1 | |||
| BX099841.1 |

Genomic Map