Assay Details
Target Gene Details
Entrez Gene ID: | 378708 |
Gene Name: | apoptosis-inducing, TAF9-like domain 1 |
Gene Aliases: |
CENP-S, CENPS, FAAP16, MHF1 |
Location: |
Chr.1:10430102-10442815 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APITD1 | NM_199294.2 | NP_954988.1 | ||
NR_036462.1 | ||||
BC029430.1 | AAH29430.1 | |||
BU902753.1 | ||||
CA311915.1 |
Target Gene Details
Entrez Gene ID: | 100526739 |
Gene Name: | APITD1-CORT readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:10430102-10452003 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APITD1-CORT | NM_001243768.1 | NP_001230697.1 | ||
NM_001270517.1 | NP_001257446.1 | |||
NM_198544.3 | NP_940946.1 | |||
NM_199006.2 | NP_950171.2 | |||
NR_037187.1 | ||||
AA858169.1 | ||||
AF516753.1 | AAM61954.1 | |||
AF521016.1 | AAM70482.1 | |||
CF127457.1 | ||||
CF128105.1 | ||||
HY054077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2764192 | Chr.1:10323676 - 10534129 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2758920 | Chr.1:10178012 - 10522497 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
