Assay Details
Target Gene Details
Entrez Gene ID: | 205 |
Gene Name: | adenylate kinase 4 |
Gene Aliases: |
AK 4, AK3, AK3L1, AK3L2 |
Location: |
Chr.1:65147342-65232145 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK4 | NM_001005353.2 | NP_001005353.1 | ||
| NM_013410.3 | NP_037542.1 | |||
| NM_203464.2 | NP_982289.1 | |||
| XM_017000612.1 | XP_016856101.1 | |||
| XM_017000613.1 | XP_016856102.1 | |||
| AK025926.1 | ||||
| AK309057.1 | ||||
| AK313611.1 | ||||
| BC016180.1 | AAH16180.1 | |||
| BC040224.1 | AAH40224.1 | |||
| BC066944.1 | AAH66944.1 | |||
| BC136886.1 | ||||
| BC146653.1 | ||||
| BC148270.1 | ||||
| BG703687.1 | ||||
| BM763432.1 | ||||
| BU164554.1 | ||||
| CR456830.1 | CAG33111.1 | |||
| DB061266.1 | ||||
| DR003868.1 | ||||
| X60673.1 | CAA43088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv173n100 | Chr.1:65102651 - 65215491 on Build GRCh38 | Gain |
|
| nsv945982 | Chr.1:65152931 - 65154711 on Build GRCh38 | Duplication |
|
| esv2676423 | Chr.1:64977641 - 65405798 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map