Assay Details
Target Gene Details
Entrez Gene ID: | 23463 |
Gene Name: | isoprenylcysteine carboxyl methyltransferase |
Gene Aliases: |
HSTE14, MST098, MSTP098, PCCMT, PCMT, PPMT |
Location: |
Chr.1:6221193-6235984 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ICMT | NM_012405.3 | NP_036537.1 | ||
| XM_011541140.2 | XP_011539442.1 | |||
| AF064084.1 | AAC16554.1 | |||
| AK092890.1 | ||||
| BC028168.1 | AAH28168.1 | |||
| CR541671.1 | CAG46472.1 | |||
| CR541711.1 | CAG46512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8n67 | Chr.1:6220621 - 6264303 on Build GRCh38 | Gain |
|
| nsv831425 | Chr.1:6129291 - 6264418 on Build GRCh38 | Loss |
|
| nsv545236 | Chr.1:6161915 - 6262876 on Build GRCh38 | Loss |
|
| nsv508825 | Chr.1:6221275 - 6286549 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map