Assay Details
Target Gene Details
Entrez Gene ID: | 199990 |
Gene Name: | Fanconi anemia core complex associated protein 20 |
Gene Aliases: |
C1orf86, FP7162 |
Location: |
Chr.1:2184460-2212720 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAAP20 | NM_001146310.1 | NP_001139782.1 | ||
| NM_001282670.1 | NP_001269599.1 | |||
| XM_006710419.3 | XP_006710482.1 | |||
| XM_011540914.2 | XP_011539216.1 | |||
| XM_011540921.2 | XP_011539223.1 | |||
| XM_017000553.1 | XP_016856042.1 | |||
| AF461903.1 | AAQ04817.1 | |||
| AK126870.1 | BAC86730.1 | |||
| AK127994.1 | BAC87220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss |
|
| dgv49n54 | Chr.1:2151050 - 2289783 on Build GRCh38 | Loss |
|
| nsv951530 | Chr.1:2153662 - 2229961 on Build GRCh38 | Deletion |
|
| nsv470682 | Chr.1:2188394 - 2378754 on Build GRCh38 | Loss |
|
| nsv834557 | Chr.1:2117631 - 2311104 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142571156] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map