Assay Details
Target Gene Details
Entrez Gene ID: | 339483 |
Gene Name: | myotubularin related protein 9-like, pseudogene |
Gene Aliases: |
MTMR9L |
Location: |
Chr.1:32231660-32241710 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTMR9LP | NR_026850.1 | |||
| AK055560.1 | ||||
| AK096109.1 | ||||
| AL833686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv300 | Chr.1:32206912 - 32241689 on Build GRCh38 | Insertion |
|

Genomic Map