Assay Details
Target Gene Details
Entrez Gene ID: | 50651 |
Gene Name: | solute carrier family 45 member 1 |
Gene Aliases: |
DNB5 |
Location: |
Chr.1:8318085-8344167 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC45A1 | NM_001080397.2 | NP_001073866.2 | ||
| XM_011541530.1 | XP_011539832.1 | |||
| XM_011541531.1 | XP_011539833.1 | |||
| AF118274.1 | 1 | 89 | AAD27583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012977 | Chr.1:8252522 - 8485210 on Build GRCh38 | Loss |
|
| nsv1743 | Chr.1:8317693 - 8351656 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map