Assay Details
Target Gene Details
Entrez Gene ID: | 64123 |
Gene Name: | adhesion G protein-coupled receptor L4 |
Gene Aliases: |
ELTD1, ETL, KPG_003 |
Location: |
Chr.1:78889764-79006810 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL4 | NM_022159.3 | NP_071442.2 | ||
| AB032017.1 | BAD83584.1 | |||
| AF192403.1 | AAG33021.1 | |||
| AK129766.1 | ||||
| AK172750.1 | BAD18736.1 | |||
| AK292894.1 | ||||
| AK313738.1 | ||||
| AY358360.1 | AAQ88726.1 | |||
| BC025721.1 | AAH25721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001291 | Chr.1:78598959 - 79684864 on Build GRCh38 | Loss |
|
| nsv526221 | Chr.1:78731939 - 79021395 on Build GRCh38 | Loss |
|
| nsv1010441 | Chr.1:78854614 - 79089630 on Build GRCh38 | Loss |
|
| nsv546619 | Chr.1:78767808 - 79149304 on Build GRCh38 | Loss |
|
| dgv331n54 | Chr.1:78893142 - 78954806 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77198934] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map