Assay Details
Target Gene Details
Entrez Gene ID: | 84879 |
Gene Name: | major facilitator superfamily domain containing 2A |
Gene Aliases: |
MCPH15, MFSD2, NLS1 |
Location: |
Chr.1:39955112-39969968 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD2A | NM_001136493.2 | NP_001129965.1 | ||
| NM_001287808.1 | NP_001274737.1 | |||
| NM_001287809.1 | NP_001274738.1 | |||
| NM_032793.4 | NP_116182.2 | |||
| NR_109896.1 | ||||
| XM_005271285.1 | XP_005271342.1 | |||
| XM_011542312.2 | XP_011540614.1 | |||
| AB073383.1 | BAD38634.1 | |||
| AF289609.1 | AAL55793.1 | |||
| AK027396.1 | 1 | 969 | ||
| AK074186.1 | ||||
| AK075183.1 | BAC11456.1 | |||
| AK093223.1 | BAC04100.1 | |||
| AK291540.1 | ||||
| AK297991.1 | ||||
| AK301533.1 | ||||
| AL834184.1 | ||||
| AY358636.1 | AAQ88999.1 | |||
| BC011587.1 | AAH11587.1 | |||
| BC092414.1 | AAH92414.1 | |||
| BP321361.1 | ||||
| DC297497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map