Assay Details
Target Gene Details
Entrez Gene ID: | 11313 |
Gene Name: | lysophospholipase II |
Gene Aliases: |
APT-2, APT2, DJ886K2.4 |
Location: |
Chr.1:23790789-23795539 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYPLA2 | NM_007260.2 | NP_009191.1 | ||
| XM_005245728.4 | XP_005245785.1 | |||
| XM_017000165.1 | XP_016855654.1 | |||
| AF098668.1 | AAC72844.1 | |||
| BC017034.2 | AAH17034.1 | |||
| BC017193.2 | AAH17193.1 |
More Information
Set Membership: |
|

Genomic Map