Assay Details
Target Gene Details
Entrez Gene ID: | 1953 |
Gene Name: | multiple EGF like domains 6 |
Gene Aliases: |
EGFL3 |
Location: |
Chr.1:3487942-3624757 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 32 - Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEGF6 | NM_001409.3 | NP_001400.3 | ||
| XM_006710406.3 | XP_006710469.1 | |||
| XM_011540885.1 | XP_011539187.1 | |||
| XM_011540886.1 | XP_011539188.1 | |||
| XM_011540887.2 | XP_011539189.1 | |||
| XM_011540888.2 | XP_011539190.1 | |||
| XM_017000533.1 | XP_016856022.1 | |||
| AB011539.2 | BAA32467.2 | |||
| AB231860.1 | ||||
| BU501408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758915 | Chr.1:3372374 - 3632289 on Build GRCh38 | Gain |
|
| nsv470687 | Chr.1:3356499 - 3763210 on Build GRCh38 | Loss |
|
| nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain |
|
| nsv951959 | Chr.1:3051037 - 3812036 on Build GRCh38 | Deletion |
|
| esv3893545 | Chr.1:3460878 - 3512044 on Build GRCh38 | Loss |
|
| nsv545125 | Chr.1:3490443 - 3540731 on Build GRCh38 | Loss |
|
| nsv830203 | Chr.1:3446374 - 3607752 on Build GRCh38 | Loss |
|
| nsv299 | Chr.1:3476928 - 3512705 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs149711898] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map