Assay Details
Target Gene Details
Entrez Gene ID: | 100130557 |
Gene Name: | NFYC antisense RNA 1 |
Gene Aliases: |
0808y08y |
Location: |
Chr.1:40689080-40692261 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYC-AS1 | NR_024567.1 | 1 | 1530 | |
| AK094323.1 | 1 | 1335 | BAC04330.1 |
Target Gene Details
Entrez Gene ID: | 9783 |
Gene Name: | regulating synaptic membrane exocytosis 3 |
Gene Aliases: |
NIM3, RIM3 |
Location: |
Chr.1:40620680-40692104 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIMS3 | XM_011542479.2 | XP_011540781.1 |
More Information
Set Membership: |
|

Genomic Map