Assay Details
Target Gene Details
Entrez Gene ID: | 51060 |
Gene Name: | thioredoxin domain containing 12 |
Gene Aliases: |
AG1, AGR1, ERP16, ERP18, ERP19, PDIA16, TLP19, hAG-1, hTLP19 |
Location: |
Chr.1:52020131-52056171 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNDC12 | NM_015913.3 | NP_056997.1 | ||
| AF131758.1 | AAD20035.1 | |||
| AF543416.1 | AAN34781.1 | |||
| AK056169.1 | ||||
| AK075409.1 | ||||
| AY358982.1 | AAQ89341.1 | |||
| BC001493.2 | AAH01493.1 | |||
| BC008913.2 | AAH08913.1 | |||
| BC008953.2 | AAH08953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004103 | Chr.1:52030594 - 52087736 on Build GRCh38 | Loss |
|
| nsv829848 | Chr.1:51902939 - 52076027 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map