Assay Details
Target Gene Details
Entrez Gene ID: | 100506504 |
Gene Name: | uncharacterized LOC100506504 |
Gene Aliases: |
- |
Location: |
Chr.1:2181811-2184176 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506504 | XM_011542503.2 | 2 | 1266 | XP_011540805.1 |
Target Gene Details
Entrez Gene ID: | 5590 |
Gene Name: | protein kinase C zeta |
Gene Aliases: |
PKC-ZETA, PKC2 |
Location: |
Chr.1:2050470-2185395 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCZ | NM_001033581.1 | NP_001028753.1 | ||
| NM_001033582.1 | NP_001028754.1 | |||
| NM_001242874.1 | NP_001229803.1 | |||
| NM_002744.4 | NP_002735.3 | |||
| XM_017001790.1 | XP_016857279.1 | |||
| XM_017001791.1 | XP_016857280.1 | |||
| XM_017001796.1 | XP_016857285.1 | |||
| XM_017001798.1 | XP_016857287.1 | |||
| XM_017001799.1 | XP_016857288.1 | |||
| XM_017001801.1 | XP_016857290.1 | |||
| XM_017001804.1 | XP_016857293.1 | |||
| XM_017001805.1 | XP_016857294.1 | |||
| AK097627.1 | ||||
| AK131526.1 | ||||
| AK290995.1 | ||||
| AK294649.1 | ||||
| AK294782.1 | ||||
| AK302160.1 | ||||
| BC008058.2 | AAH08058.1 | |||
| BC014270.2 | AAH14270.1 | |||
| BT007082.1 | AAP35745.1 | |||
| L14283.1 | AAA36488.1 | |||
| Z15108.1 | CAA78813.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss |
|
| dgv49n54 | Chr.1:2151050 - 2289783 on Build GRCh38 | Loss |
|
| nsv951530 | Chr.1:2153662 - 2229961 on Build GRCh38 | Deletion |
|
| nsv470681 | Chr.1:2092625 - 2182126 on Build GRCh38 | Loss |
|
| nsv834557 | Chr.1:2117631 - 2311104 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map