Assay Details
Target Gene Details
Entrez Gene ID: | 22996 |
Gene Name: | tetratricopeptide repeat domain 39A |
Gene Aliases: |
C1orf34, DEME-6 |
Location: |
Chr.1:51287258-51345116 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
More Information
Set Membership: |
|

Genomic Map