Assay Details
Target Gene Details
Entrez Gene ID: | 91156 |
Gene Name: | immunoglobulin-like and fibronectin type III domain containing 1 |
Gene Aliases: |
EEF1A2BP1 |
Location: |
Chr.1:201190780-201228952 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGFN1 | NM_001164586.1 | NP_001158058.1 | ||
| XM_005245580.3 | XP_005245637.1 | |||
| XM_006711618.3 | XP_006711681.1 | |||
| XM_011510120.2 | XP_011508422.1 | |||
| XM_011510121.2 | XP_011508423.1 | |||
| XM_017002787.1 | XP_016858276.1 |
More Information
Set Membership: |
|

Genomic Map