Assay Details
Target Gene Details
Entrez Gene ID: | 23127 |
Gene Name: | collagen beta(1-O)galactosyltransferase 2 |
Gene Aliases: |
C1orf17, GLT25D2 |
Location: |
Chr.1:183929662-184051612 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COLGALT2 | NM_001303420.1 | NP_001290349.1 | ||
| AK293973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525878 | Chr.1:183873970 - 183944406 on Build GRCh38 | Gain |
|
| nsv832026 | Chr.1:183815931 - 183992713 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111306887] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map