Assay Details
Target Gene Details
Entrez Gene ID: | 23358 |
Gene Name: | ubiquitin specific peptidase 24 |
Gene Aliases: |
- |
Location: |
Chr.1:55066359-55215374 on Build GRCh38 |
Assay Gene Location: | Within Exon 72 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP24 | NM_015306.2 | 68 | 8128 | NP_056121.2 |
| XM_005270690.3 | 55 | 6169 | XP_005270747.1 | |
| XM_017000831.1 | 69 | 8193 | XP_016856320.1 | |
| XM_017000832.1 | 69 | 8190 | XP_016856321.1 | |
| XM_017000833.1 | 69 | 7685 | XP_016856322.1 | |
| XM_017000834.1 | 68 | 7625 | XP_016856323.1 | |
| AB028980.1 | 27 | 2947 | BAA83009.1 | |
| AK000321.1 | 12 | 1112 | BAA91084.1 | |
| AL832370.1 | 30 | 3354 | ||
| BC029660.1 | 13 | 1181 | AAH29660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518539 | Chr.1:55034756 - 55087881 on Build GRCh38 | Loss |
|
| esv3577877 | Chr.1:55068894 - 55073692 on Build GRCh38 | Loss |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map