Assay Details
Target Gene Details
Entrez Gene ID: | 148979 |
Gene Name: | GLIS family zinc finger 1 |
Gene Aliases: |
- |
Location: |
Chr.1:53506233-53739171 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLIS1 | NM_147193.2 | NP_671726.2 | ||
| XM_017000408.1 | XP_016855897.1 | |||
| XM_017000409.1 | XP_016855898.1 | |||
| XM_017000410.1 | XP_016855899.1 | |||
| XM_017000411.1 | XP_016855900.1 | |||
| AK090634.1 | BAC03494.1 | |||
| AK093474.1 | BAC04178.1 | |||
| BC101799.1 | ||||
| BC104911.1 | AAI04912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522165 | Chr.1:53506376 - 53539387 on Build GRCh38 | Loss |
|
| nsv509269 | Chr.1:53296636 - 53583096 on Build GRCh38 | Insertion |
|
| esv2758936 | Chr.1:53452879 - 53563563 on Build GRCh38 | Loss |
|
| nsv829882 | Chr.1:53449992 - 53629940 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map