Assay Details
Target Gene Details
Entrez Gene ID: | 5768 |
Gene Name: | quiescin sulfhydryl oxidase 1 |
Gene Aliases: |
Q6, QSCN6 |
Location: |
Chr.1:180154833-180198034 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QSOX1 | NM_001004128.2 | NP_001004128.1 | ||
| NM_002826.4 | NP_002817.2 | |||
| AB209280.1 | BAD92517.1 | |||
| AF361868.1 | AAM00263.1 | |||
| AK290842.1 | ||||
| AK290887.1 | ||||
| AY358941.1 | AAQ89300.1 | |||
| BC017692.1 | AAH17692.1 | |||
| BC100023.1 | AAI00024.1 | |||
| BX382719.2 | ||||
| L42379.1 | AAA89173.1 | |||
| U97276.2 | AAC09010.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001565 | Chr.1:180167906 - 180201389 on Build GRCh38 | Loss |
|
| dgv475n100 | Chr.1:180156531 - 180210921 on Build GRCh38 | Gain |
|
| dgv476n100 | Chr.1:180157303 - 180231969 on Build GRCh38 | Gain |
|
| nsv832004 | Chr.1:180159367 - 180366405 on Build GRCh38 | Gain |
|
| dgv474n100 | Chr.1:180152345 - 180273524 on Build GRCh38 | Gain |
|
| nsv952285 | Chr.1:180164566 - 180200265 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map