Assay Details
Target Gene Details
Entrez Gene ID: | 10420 |
Gene Name: | testis-specific kinase 2 |
Gene Aliases: |
- |
Location: |
Chr.1:45343883-45491342 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TESK2 | NM_001320800.1 | NP_001307729.1 | ||
| NM_007170.2 | NP_009101.2 | |||
| XM_005270355.3 | XP_005270412.1 | |||
| XM_006710287.2 | XP_006710350.1 | |||
| XM_011540497.1 | XP_011538799.1 | |||
| XM_011540500.2 | XP_011538802.1 | |||
| XM_017000047.1 | XP_016855536.1 | |||
| XM_017000048.1 | XP_016855537.1 | |||
| XM_017000049.1 | XP_016855538.1 | |||
| AB057597.1 | BAB62909.1 | |||
| AJ132545.1 | CAB41970.1 | |||
| AK027573.1 | ||||
| AK294172.1 | ||||
| BC033085.1 | AAH33085.1 | |||
| BX648441.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750837 | Chr.1:45330443 - 45442591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map