Assay Details
Target Gene Details
Entrez Gene ID: | 100287898 |
Gene Name: | tetratricopeptide repeat domain 34 |
Gene Aliases: |
- |
Location: |
Chr.1:2636988-2789737 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC34 | NM_001242672.1 | NP_001229601.1 | ||
| XM_016999990.1 | XP_016855479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv21805 | Chr.1:2634485 - 2702743 on Build GRCh38 | Gain+Loss |
|
| nsv824586 | Chr.1:2649029 - 2652002 on Build GRCh38 | Gain |
|
| esv994553 | Chr.1:2641028 - 2652640 on Build GRCh38 | Insertion |
|
| nsv470684 | Chr.1:2394570 - 2930923 on Build GRCh38 | Loss |
|
| nsv428399 | Chr.1:2622539 - 3235926 on Build GRCh38 | Gain |
|
| nsv545083 | Chr.1:2575976 - 2653099 on Build GRCh38 | Gain |
|

Genomic Map