Assay Details
Target Gene Details
Entrez Gene ID: | 11146 |
Gene Name: | glomulin, FKBP associated protein |
Gene Aliases: |
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM |
Location: |
Chr.1:92245786-92441936 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLMN | NM_001319683.1 | NP_001306612.1 | ||
| NM_053274.2 | NP_444504.1 | |||
| NR_135089.1 | ||||
| XM_005270401.3 | XP_005270458.1 | |||
| XM_006710309.2 | XP_006710372.1 | |||
| XM_011540546.2 | XP_011538848.1 | |||
| XM_017000137.1 | XP_016855626.1 | |||
| XM_017000138.1 | XP_016855627.1 | |||
| XM_017000139.1 | XP_016855628.1 | |||
| XM_017000140.1 | XP_016855629.1 | |||
| XM_017000141.1 | XP_016855630.1 | |||
| XM_017000142.1 | XP_016855631.1 | |||
| XM_017000143.1 | XP_016855632.1 | |||
| XM_017000144.1 | XP_016855633.1 | |||
| AJ302735.1 | CAC82938.1 | |||
| AJ347709.1 | CAC69882.1 | |||
| AK295971.1 | ||||
| BC001257.1 | AAH01257.1 | |||
| U73704.1 | AAC50908.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830592 | Chr.1:92203725 - 92340072 on Build GRCh38 | Gain |
|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map