Assay Details
Target Gene Details
Entrez Gene ID: | 729614 |
Gene Name: | uncharacterized LOC729614 |
Gene Aliases: |
- |
Location: |
Chr.1:15834215-15848147 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ37453 | NR_024279.1 | 2 | 2548 | |
| AK094772.1 | 1 | 1462 | BAC04420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524044 | Chr.1:15675093 - 15835341 on Build GRCh38 | Loss |
|
| nsv1011969 | Chr.1:15826990 - 15875045 on Build GRCh38 | Gain+Loss |
|
| dgv154n54 | Chr.1:15827003 - 15834894 on Build GRCh38 | Loss |
|
| nsv833647 | Chr.1:15794042 - 15952334 on Build GRCh38 | Loss |
|
| nsv545619 | Chr.1:15802946 - 15834894 on Build GRCh38 | Loss |
|
| nsv511126 | Chr.1:15822806 - 15835341 on Build GRCh38 | Loss |
|
| dgv37n100 | Chr.1:15823408 - 15924038 on Build GRCh38 | Gain |
|
| esv2764241 | Chr.1:15827002 - 15855150 on Build GRCh38 | Loss |
|
| nsv3409 | Chr.1:15809988 - 15852694 on Build GRCh38 | Deletion |
|

Genomic Map