Assay Details
Target Gene Details
Entrez Gene ID: | 343052 |
Gene Name: | immunoglobulin superfamily DCC subclass member 3 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.1:153793947-153796302 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC343052 | NR_126565.1 | |||
| BX118114.1 | ||||
| CN292333.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515962 | Chr.1:153779663 - 153800953 on Build GRCh38 | Loss |
|
| nsv547941 | Chr.1:153736430 - 153828622 on Build GRCh38 | Loss |
|

Genomic Map