Assay Details
Target Gene Details
Entrez Gene ID: | 149069 |
Gene Name: | doublecortin domain containing 2B |
Gene Aliases: |
- |
Location: |
Chr.1:32208813-32216196 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCDC2B | NM_001099434.1 | NP_001092904.1 | ||
| XM_017000413.1 | XP_016855902.1 | |||
| XM_017000414.1 | XP_016855903.1 | |||
| BC128073.1 |
Target Gene Details
Entrez Gene ID: | 56063 |
Gene Name: | transmembrane protein 234 |
Gene Aliases: |
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7 |
Location: |
Chr.1:32204807-32222371 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM234 | XM_011541795.2 | XP_011540097.1 | ||
| XM_017001814.1 | XP_016857303.1 | |||
| XM_017001815.1 | XP_016857304.1 | |||
| XM_017001817.1 | XP_016857306.1 | |||
| XM_017001818.1 | XP_016857307.1 | |||
| XM_017001819.1 | XP_016857308.1 | |||
| XM_017001821.1 | XP_016857310.1 | |||
| XM_017001827.1 | XP_016857316.1 | |||
| XM_017001828.1 | XP_016857317.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv300 | Chr.1:32206912 - 32241689 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map