Assay Details
Target Gene Details
Entrez Gene ID: | 200150 |
Gene Name: | phospholipase D family member 5 |
Gene Aliases: |
PLDC |
Location: |
Chr.1:242085051-242530546 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLD5 | XM_011544119.2 | 7 | 3733 | XP_011542421.2 |
| XM_011544120.2 | 10 | 3378 | XP_011542422.1 | |
| XM_011544121.2 | 7 | 2867 | XP_011542423.1 | |
| XM_011544122.2 | 7 | 4328 | XP_011542424.1 | |
| XM_017000568.1 | 10 | 3360 | XP_016856057.1 | |
| XM_017000569.1 | 10 | 5339 | XP_016856058.1 | |
| XM_017000570.1 | 8 | 4079 | XP_016856059.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589364 | Chr.1:242077013 - 242088139 on Build GRCh38 | Loss |
|
| dgv16e55 | Chr.1:241861796 - 242169657 on Build GRCh38 | Gain |
|
| nsv470788 | Chr.1:242037945 - 242392647 on Build GRCh38 | Gain |
|
| nsv528016 | Chr.1:242060097 - 242288048 on Build GRCh38 | Gain |
|
| nsv1002309 | Chr.1:241878940 - 242132333 on Build GRCh38 | Gain |
|
| nsv508703 | Chr.1:241983638 - 242088799 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map