Assay Details
Target Gene Details
Entrez Gene ID: | 7804 |
Gene Name: | LDL receptor related protein 8 |
Gene Aliases: |
APOER2, HSZ75190, LRP-8, MCI1 |
Location: |
Chr.1:53242364-53328149 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRP8 | NM_001018054.2 | 18 | 5042 | NP_001018064.1 |
| NM_004631.4 | 19 | 5219 | NP_004622.2 | |
| NM_017522.4 | 16 | 4430 | NP_059992.3 | |
| NM_033300.3 | 17 | 4709 | NP_150643.2 | |
| XM_005271173.3 | 20 | 5168 | XP_005271230.1 | |
| XM_006710881.3 | 19 | 4991 | XP_006710944.1 | |
| XM_006710882.3 | 19 | 4943 | XP_006710945.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011750 | Chr.1:53226981 - 53261502 on Build GRCh38 | Gain |
|
| esv2422213 | Chr.1:53179891 - 53258767 on Build GRCh38 | Duplication |
|
| nsv829871 | Chr.1:53200871 - 53383048 on Build GRCh38 | Loss |
|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117431410] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map