Assay Details
Target Gene Details
Entrez Gene ID: | 91156 |
Gene Name: | immunoglobulin-like and fibronectin type III domain containing 1 |
Gene Aliases: |
EEF1A2BP1 |
Location: |
Chr.1:201190780-201228952 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGFN1 | NM_001164586.1 | NP_001158058.1 | ||
| XM_005245580.3 | XP_005245637.1 | |||
| XM_006711618.3 | XP_006711681.1 | |||
| XM_017002787.1 | XP_016858276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428301 | Chr.1:201059216 - 201191346 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map