Assay Details
Target Gene Details
Entrez Gene ID: | 58512 |
Gene Name: | DLG associated protein 3 |
Gene Aliases: |
DAP3, SAPAP3 |
Location: |
Chr.1:34862120-34929585 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP3 | NM_001080418.2 | 6 | 1805 | NP_001073887.1 |
| XM_011541879.2 | 7 | 3690 | XP_011540181.1 | |
| XM_011541880.2 | 2 | 1514 | XP_011540182.1 | |
| BC114370.1 |
More Information
Set Membership: |
|

Genomic Map