Assay Details
Target Gene Details
Entrez Gene ID: | 58512 |
Gene Name: | DLG associated protein 3 |
Gene Aliases: |
DAP3, SAPAP3 |
Location: |
Chr.1:34862120-34929585 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP3 | NM_001080418.2 | NP_001073887.1 | ||
| XM_011541879.2 | XP_011540181.1 | |||
| XM_011541880.2 | XP_011540182.1 | |||
| AF131778.1 | AAD20042.1 | |||
| BC114370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012995 | Chr.1:34786068 - 34884653 on Build GRCh38 | Gain |
|
| esv2670062 | Chr.1:34883655 - 34884589 on Build GRCh38 | Deletion |
|
| esv3585712 | Chr.1:34883656 - 34884595 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map