Assay Details
Target Gene Details
Entrez Gene ID: | 388630 |
Gene Name: | TraB domain containing 2B |
Gene Aliases: |
TIKI2 |
Location: |
Chr.1:47760528-47997367 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRABD2B | NM_001194986.1 | NP_001181915.1 | ||
| XM_006710640.3 | XP_006710703.1 | |||
| XM_011541443.2 | XP_011539745.1 | |||
| XM_011541444.2 | XP_011539746.1 | |||
| XM_011541445.2 | XP_011539747.1 | |||
| XM_017001260.1 | XP_016856749.1 | |||
| XM_017001261.1 | XP_016856750.1 | |||
| XM_017001262.1 | XP_016856751.1 | |||
| XM_017001263.1 | XP_016856752.1 | |||
| JQ653416.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829760 | Chr.1:47645454 - 47842516 on Build GRCh38 | Loss |
|
| nsv1001126 | Chr.1:47698950 - 47855190 on Build GRCh38 | Gain |
|
| nsv1003161 | Chr.1:47742043 - 47820891 on Build GRCh38 | Loss |
|
| nsv461450 | Chr.1:47771291 - 47816912 on Build GRCh38 | Loss |
|
| nsv998583 | Chr.1:47766459 - 47851205 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map