Assay Details
Target Gene Details
Entrez Gene ID: | 5538 |
Gene Name: | palmitoyl-protein thioesterase 1 |
Gene Aliases: |
CLN1, INCL, PPT |
Location: |
Chr.1:40072710-40097470 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPT1 | NM_000310.3 | NP_000301.1 | ||
| NM_001142604.1 | NP_001136076.1 | |||
| XM_005271008.1 | XP_005271065.1 | |||
| AK075496.1 | ||||
| AK301682.1 | ||||
| AK302232.1 | ||||
| AK310799.1 | ||||
| AK312287.1 | ||||
| BC008426.1 | AAH08426.1 | |||
| CR542053.1 | CAG46850.1 | |||
| U44772.1 | AAB06236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |
|
| nsv10128 | Chr.1:40082338 - 40088067 on Build GRCh38 | Loss |
|
| esv2747330 | Chr.1:40086621 - 40087965 on Build GRCh38 | Deletion |
|
| esv6823 | Chr.1:40086476 - 40088121 on Build GRCh38 | Loss |
|
| esv3585798 | Chr.1:40086684 - 40087956 on Build GRCh38 | Loss |
|
| nsv829660 | Chr.1:40073032 - 40266408 on Build GRCh38 | Gain+Loss |
|
| esv2663819 | Chr.1:40086685 - 40087968 on Build GRCh38 | Deletion |
|
| esv2750836 | Chr.1:40038107 - 40093576 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map