Assay Details
Target Gene Details
Entrez Gene ID: | 26289 |
Gene Name: | adenylate kinase 5 |
Gene Aliases: |
AK6 |
Location: |
Chr.1:77281908-77559969 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK5 | NM_012093.3 | NP_036225.2 | ||
| NM_174858.2 | NP_777283.1 | |||
| XM_005270739.4 | XP_005270796.1 | |||
| XM_006710572.3 | XP_006710635.1 | |||
| XM_017001008.1 | XP_016856497.1 | |||
| XM_017001009.1 | XP_016856498.1 | |||
| XM_017001010.1 | XP_016856499.1 | |||
| XM_017001011.1 | XP_016856500.1 | |||
| XM_017001012.1 | XP_016856501.1 | |||
| AF445193.1 | AAP97322.1 | |||
| AK090967.1 | BAC03558.1 | |||
| AY171600.2 | AAO16520.2 | |||
| BC033896.1 | AAH33896.1 | |||
| BC036666.1 | AAH36666.1 | |||
| DN993295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758945 | Chr.1:77309329 - 77322777 on Build GRCh38 | Loss |
|
| nsv818234 | Chr.1:77308728 - 77312923 on Build GRCh38 | Loss |
|
| esv3586520 | Chr.1:77308819 - 77315915 on Build GRCh38 | Loss |
|
| esv2676174 | Chr.1:77308818 - 77315904 on Build GRCh38 | Deletion |
|
| nsv441697 | Chr.1:77309187 - 77314147 on Build GRCh38 | Loss |
|
| esv22538 | Chr.1:77309059 - 77315769 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map