Assay Details
Target Gene Details
Entrez Gene ID: | 5192 |
Gene Name: | peroxisomal biogenesis factor 10 |
Gene Aliases: |
NALD, PBD6A, PBD6B, RNF69 |
Location: |
Chr.1:2404802-2412622 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEX10 | NM_002617.3 | NP_002608.1 | ||
| NM_153818.1 | NP_722540.1 | |||
| XM_011541573.1 | XP_011539875.1 | |||
| XM_011541576.1 | XP_011539878.1 | |||
| AB013818.1 | BAA87895.1 | |||
| AF060502.1 | AAC18133.1 | |||
| AK124816.1 | ||||
| BC000543.2 | AAH00543.1 | |||
| BC018198.1 | AAH18198.1 | |||
| CR542077.1 | CAG46874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951557 | Chr.1:2266062 - 2564661 on Build GRCh38 | Deletion |
|
| nsv545072 | Chr.1:2401592 - 2475448 on Build GRCh38 | Loss |
|
| nsv470684 | Chr.1:2394570 - 2930923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map