Assay Details
Target Gene Details
Entrez Gene ID: | 151258 |
Gene Name: | solute carrier family 38 member 11 |
Gene Aliases: |
AVT2 |
Location: |
Chr.2:164897745-164955721 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC38A11 | XM_005246350.4 | 12 | 2290 | XP_005246407.1 |
| XM_006712337.3 | 11 | 2286 | XP_006712400.1 | |
| XM_011510737.2 | 11 | 2382 | XP_011509039.1 | |
| XM_011510742.2 | 9 | 2078 | XP_011509044.1 | |
| XM_017003455.1 | 12 | 2526 | XP_016858944.1 | |
| XM_017003456.1 | 11 | 2460 | XP_016858945.1 | |
| XM_017003457.1 | 11 | 2451 | XP_016858946.1 | |
| XM_017003458.1 | 11 | 2411 | XP_016858947.1 | |
| XM_017003459.1 | 10 | 2149 | XP_016858948.1 | |
| XM_017003463.1 | 11 | 1923 | XP_016858952.1 | |
| XM_017003464.1 | 8 | 2135 | XP_016858953.1 | |
| XM_017003465.1 | 7 | 1663 | XP_016858954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834444 | Chr.2:164865781 - 165038205 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map