Assay Details
Target Gene Details
Entrez Gene ID: | 10617 |
Gene Name: | STAM binding protein |
Gene Aliases: |
AMSH, MICCAP |
Location: |
Chr.2:73828911-73873656 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAMBP | NM_006463.4 | NP_006454.1 | ||
| NM_201647.2 | NP_964010.1 | |||
| NM_213622.2 | NP_998787.1 | |||
| XM_005264088.3 | XP_005264145.1 | |||
| XM_011532483.2 | XP_011530785.1 | |||
| XM_011532484.2 | XP_011530786.1 | |||
| XM_011532485.2 | XP_011530787.1 | |||
| XM_017003175.1 | XP_016858664.1 | |||
| XM_017003176.1 | XP_016858665.1 | |||
| XM_017003177.1 | XP_016858666.1 | |||
| XM_017003178.1 | XP_016858667.1 | |||
| XM_017003179.1 | XP_016858668.1 | |||
| XM_017003180.1 | XP_016858669.1 | |||
| XM_017003181.1 | XP_016858670.1 | |||
| AF052135.1 | ||||
| BC007682.1 | AAH07682.1 | |||
| BC065574.1 | AAH65574.1 | |||
| BC101467.1 | AAI01468.1 | |||
| BC101469.1 | AAI01470.1 | |||
| BM478027.1 | ||||
| BQ929694.1 | ||||
| DN919763.1 | ||||
| DR158757.1 | ||||
| EU927390.1 | ||||
| U73522.1 | AAD05037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834260 | Chr.2:73678979 - 73849393 on Build GRCh38 | Gain |
|
| esv2759060 | Chr.2:73489905 - 73851465 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map