Assay Details
Target Gene Details
Entrez Gene ID: | 91543 |
Gene Name: | radical S-adenosyl methionine domain containing 2 |
Gene Aliases: |
2510004L01Rik, cig33, cig5, vig1 |
Location: |
Chr.2:6865733-6898239 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSAD2 | XM_011510414.2 | XP_011508716.1 | ||
| XM_011510415.2 | XP_011508717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833326 | Chr.2:6844896 - 7002433 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map